Cytiva Frac30 Model Fraction Collector
Used as a round fraction collector. Cytiva Frac30 Model Fraction Collector allows you to collect up to 30 fractions.
Supplier: Cytiva 29023051
Description
- Controlled via the AKTA protein purification system start touch screen instrument display
- Also controlled by using UNICORN start software via computer
- Collect up to 30 fractions
- Supports four tube sizes, ranging from 1.5mL to 15mL
- Fractions can be collected in volumes ranging from 0.5mL to 15mL
- The entire fraction collector rack can be transferred as required
Specifications
Fraction Collector |