missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FR4 Rat anti-Mouse, Clone: eBioTH6, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$477.05 - $592.68

Specifications

Antigen FR4
Clone eBioTH6
Host Species Rat
Gene Alias Folate receptor 4, FOLR4 FRdelta, FRd
Applications Flow Cytometry,Functional Assay,Immunoprecipitation
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014563 Life Technologies
LS16544682
100μg Each for $477.05
5014564 Life Technologies
LS16544685
500μg Each for $592.68
Specifications

Description

The monoclonal antibody eBioTh6 recognizes FR4, also known as FR δ and FBP3 (folate binding protein3). FR4 is a heavily glycosylated 35 kD receptor for folic acid and the physiologic circulating form of the vitamin, N5-methyltetrahydrofolate. Natural T regs have high levels of FR4 and together with CD25 can be used to distinguish natural Tregs, effector T cells, memory-like T cells and naïve T cells. In vivo studies with eBioTH6 show a decrease in the number of CD4+CD25+ cells and an enhanced tumor response leading to the elimination of advanced stages of tumors.

Based on co-staining studies, the epitopes recognized by eBioTH6 and eBio12A5 (cat 51-5445) are different thereby allowing functional studies with TH6 to be evaluated with eBio12A5.
Description & Specifications

Specifications

FR4
Rat
Flow Cytometry,Functional Assay,Immunoprecipitation
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL
eBioTH6
Folate receptor 4, FOLR4 FRdelta, FRd
RUO
Purified
Primary
aqueous buffer, no sodium azide