Please login to your online account to display your discounted pricing

Tocris™ FR 139317

Highly potent, selective ETA antagonist

$283.92 - $1,179.73

Specifications

Product Type FR 139317
Inhibitors FR 139317
Molecular Weight 604.75
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

121010

 
tocris™
121010
10mg Each for $283.92

121050R

 
tocris™
121050
50mg Each for $1,179.73
Description & Specifications

Specifications

Product Type FR 139317
Inhibitors FR 139317
Molecular Weight 604.75
Purity >99

A highly potent and selective ETA endothelin receptor antagonist (Ki values are 1nM and 7.3μM at ETA and ETB subtypes respectively). Active in vivo.

  • Chemical Name: N-[N-[N-[(Hexahydro-1H-azepin-1-yl)carbonyl]-L-leucyl]-1-methyl-D-tryptophyl]-3-(2-pyridinyl)-D-alanine
  • Formula: C33H44N6O5
  • CAS Number: 142375-60-8
  • Solubility: Soluble to 100mM in 1eq. HCl and to 100mM in DMSO
  • Purity: >99