Please login to your online account to display your discounted pricing

FR 122047 hydrochloride, Tocris

Cyclooxygenase (COX-1) inhibitor

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material FR 122047 hydrochloride
Assay Percent Range >98%
Molecular Formula C23H25N3O3S·HCl
Formula Weight 459.99
Synonym 1-[[4, 5-bis(4-Methoxyphenyl)-2-thiazolyl]carbonyl]-4-methylpiperazine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

150750

 
r & d systems
RDS150750
50mg Each for $924.23

150710

 
r & d systems
RDS150710
10mg Each for $225.77
Description & Specifications

Specifications

Formula Weight 459.99
Quantity 50mg
Chemical Name or Material FR 122047 hydrochloride
Assay Percent Range >98%
Molecular Formula C23H25N3O3S·HCl
Synonym 1-[[4, 5-bis(4-Methoxyphenyl)-2-thiazolyl]carbonyl]-4-methylpiperazine hydrochloride
CAS 130717-51-0
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C