Please login to your online account to display your discounted pricing

FPR A14, Tocris

FPR agonist

$208.25 - $867.95

Chemical Identifiers

Chemical Name or Material FPR A14
Assay Percent Range >99%
Molecular Formula C23H20N2O5
Formula Weight 404.42
Synonym 1, 3-Benzodioxolane-5-carboxylic acid 4'-benzyloxy-3'-methoxybenzylidene hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

282610

 
r & d systems
RDS282610
10mg Each for $208.25

282650

 
r & d systems
RDS282650
50mg Each for $867.95
Description & Specifications

Specifications

Formula Weight 404.42
Quantity 10mg
CAS 329691-12-5
Molecular Formula C23H20N2O5
Chemical Name or Material FPR A14
Assay Percent Range >99%
Synonym 1, 3-Benzodioxolane-5-carboxylic acid 4'-benzyloxy-3'-methoxybenzylidene hydrazide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature