Please login to your online account to display your discounted pricing

FPL 64176, Tocris

Potent activator of Ca2+ channels (L-type)

$163.07 - $678.85

Chemical Identifiers

Chemical Name or Material FPL 64176
Assay Percent Range >99%
Molecular Formula C22H21NO3
Formula Weight 347.41
Synonym 2, 5-Dimethyl-4-[2-(phenylmethyl)benzoyl]-1H-pyrrole-3-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

140350

 
r & d systems
140350
50mg Each for $678.85

140310

 
r & d systems
140310
10mg Each for $163.07
Description & Specifications

Specifications

Formula Weight 347.41
Quantity 50mg
Chemical Name or Material FPL 64176
Molecular Formula C22H21NO3
Assay Percent Range >99%
Synonym 2, 5-Dimethyl-4-[2-(phenylmethyl)benzoyl]-1H-pyrrole-3-carboxylic acid methyl ester
CAS 120934-96-5
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature