missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FPA 124, Tocris

Akt/PKB inhibitor

$219.42 - $898.19

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C11H9Cl2CuN3O2S
Formula Weight 381.73
Synonym Dichloro[(2Z)-2-[(4-oxo-4H-1-benzopyran-3-yl)methylene]hydrazinecarbothioamide copper complex
Chemical Name or Material FPA 124
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
292650 R & D Systems
RDS292650
50mg Each for $898.19
292610 R & D Systems
RDS292610
10mg Each for $219.42
Specifications Description & Specifications

Chemical Identifiers

>98%
381.73
FPA 124
C11H9Cl2CuN3O2S
Dichloro[(2Z)-2-[(4-oxo-4H-1-benzopyran-3-yl)methylene]hydrazinecarbothioamide copper complex

Specifications

902779-59-3
C11H9Cl2CuN3O2S
Dichloro[(2Z)-2-[(4-oxo-4H-1-benzopyran-3-yl)methylene]hydrazinecarbothioamide copper complex
FPA 124
>98%
381.73
50mg

Safety and Handling

Recommended Storage : Store at +4°C