missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FOXP3, eFluor 660, clone: FJK-16s, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS50577382

 View more versions of this product

Catalog No. 501128851



Description

Description

FOXP3 is a member of the forkhead/winged-helix family of transcriptional regulators and is highly conserved across mammals. FOXP3 is essential for normal immune homeostasis. FOXP3 is stably and constitutively expressed at a high level in CD25 + CD4 positive regulatory T cells, a low level in CD4-positive/CD25-negative cells, and is absent in CD4-negative/CD8-positive T cells. FOXP3 may be a master regulatory gene and a more specific marker of regulatory T cells than other T cells.

Applications Tested: This FJK-16s antibody has been tested by intracellular staining and flow cytometric analysis of mouse splenocytes using the Foxp3/Transcription Factor Buffer Set (cat. 00-5523) and protocol. Please refer to intracellular Staining Protocol on Best Protocol web page - Protocol B: One-step protocol for intracellular (nuclear) proteins. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. This FJK-16s antibody has been tested by immunohistochemistry of frozen mouse tissue and can be used at less than or equal to 20 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. FOXP3 (Forkhead box protein 3) is a member of the forkhead/winged-helix family of transcriptional regulators, highly conserved across mammals, and essential for normal immune homeostasis. FOXP3 is 381 amino acids long, stably and constitutively expressed at a high level in CD25 + CD4 positive regulatory T cells, a low level in CD4-positive/CD25-negative cells, and is absent in CD4-negative/CD8-positive T cells. FOXP3 may be a master regulatory gene, and a more specific marker of regulatory T cells. Defects in the gene encoding FOXP3 protein cause the scurfy phenotype in mice. In humans FOXP3 defects play a role in IPEX syndrome (immune dysfunction, polyendocrinopathy, enteropathy, X-linked syndrome), also known as X-linked autoimmunity-allergic dysregulation (XLAAD) syndrome. Transcript variants of FOXP3 encoding different isoforms have been identified. In human breast and colon cancer cells, expression of FOXP3 is regulated by p53 in response to the DNA damage.
Specifications

Specifications

Foxp3
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q99JB6
AIID, DIETER, Forkhead box P3, foxp3, IPEX, JM2, PIDX, SCURFIN, XPID
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Bovine, Canine, Feline, Mouse, Porcine, Rat
Flow Cytometry, Immunohistochemistry (Frozen)
FJK-16s
eFluor® 660
Foxp3
Liquid
Foxp3
IgG2a, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
100037405, 20371, 317382, 444998, 491876, 506053
Documents

For Research Use Only.