Please login to your online account to display your discounted pricing

Foxp3 Rat anti-Mouse/Rat, Clone: FJK-16s, eBioscience™

Rat Monoclonal Antibody

$201.55 - $390.05

Specifications

Antigen Foxp3
Clone FJK-16s
Host Species Rat
Gene Alias Forkhead Box P3, Scurfin, JM2, Treg
Species Reactivity Bovine, Canine, cat, Mouse, Pig, Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013141

 
life technologies
LS14577380
25μg Each for $201.55

501128845

 
life technologies
LS14577382
100μg Each for $390.05
Description & Specifications

Specifications

Antigen Foxp3
Clone FJK-16s
Host Species Rat
Gene Alias Forkhead Box P3, Scurfin, JM2, Treg
Species Reactivity Bovine, Canine, cat, Mouse, Pig, Rat
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The FJK-16s antibody reacts with mouse, rat, dog, porcine, bovine and cat Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2; cross reactivity of this antibody to other proteins has not been determined. Foxp3, a 49-55kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in ‘scurfy’ (sf) mice. Constitutive high expression of foxP3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg cells), and ectopic expression of foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

Immunoblotting with FJK-16s antibody has mapped the epitope to amino acids 75-125 of the mouse Foxp3 protein. In the human, this region has been shown to be alternatively spliced at the mRNA level. Both the alternatively-spliced and non-spliced isoforms are present in the CD4+CD25+ subset of lymphocytes. Preliminary RT-PCR experiments have not revealed this alternatively-spliced isoform in mouse splenocytes, suggesting different gene regulation in the mouse and human.