Please login to your online account to display your discounted pricing

Foxp3 Rat anti-Mouse, Rat, Alexa Fluor 700, Clone: FJK-16s, eBioscience™

Rat Monoclonal Antibody

$433.55 - $564.05

Specifications

Antigen Foxp3
Clone FJK-16s
Host Species Rat
Gene Alias Forkhead Box P3, Scurfin, JM2
Species Reactivity Bovine, Canine, cat, Mouse, Pig, Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016919

 
affymetrix
56577380
25μg Each for $433.55

501128891

 
affymetrix
56577382
100μg Each for $564.05
Description & Specifications

Specifications

Antigen Foxp3
Clone FJK-16s
Host Species Rat
Gene Alias Forkhead Box P3, Scurfin, JM2
Species Reactivity Bovine, Canine, cat, Mouse, Pig, Rat
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate Alexa Fluor 700
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The FJK-16s antibody reacts with mouse, rat, dog, porcine, bovine and cat Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2; cross reactivity of this antibody to other proteins has not been determined. Foxp3, a 49-55kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in ‘scurfy’ (sf) mice. Constitutive high expression of foxP3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg cells), and ectopic expression of foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

Immunoblotting with FJK-16s antibody has mapped the epitope to amino acids 75-125 of the mouse Foxp3 protein. In the human, this region has been shown to be alternatively spliced at the mRNA level. Both the alternatively-spliced and non-spliced isoforms are present in the CD4+CD25+ subset of lymphocytes. Preliminary RT-PCR experiments have not revealed this alternatively-spliced isoform in mouse splenocytes, suggesting different gene regulation in the mouse and human.

Intracellular staining of mouse splenocytes with FJK-16s using the PE anti-mouse/rat Foxp3 Staining Set and protocol reveals approximately 2% of total cells in the C57Bl/6 strain and approximately 3-5% in the BALB/c mouse strain. Multicolor flow cytometric analysis demonstrates approximately 90% of the CD4+CD25+ cells and 4% of the CD4+CD25- cells staining with FJK-16s. B220+, CD11b+, CD11c+, and Ly6G/Gr-1+ cells do not show significant co-staining with FJK-16s.

Please see our FAQ regarding the usage of eBioscience Foxp3 reagents.

FJK-16s cross-reacts with rat, bovine, porcine and canine Foxp3. This has been demonstrated by intracellular staining of Foxp3 and flow cytometry of rat splenocytes using the same method and reagents as used for mouse tissue. Please note that FJK-16s has been optimized for use with the mouse Foxp3 Staining Set (cat. 72-5775 or 71-5775 or 77-5775). The use of other fixation and staining buffers is not recommended.