missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Foxp3 Rat anti-Human, Biotin, Clone: PCH101, eBioscience™

Rat Monoclonal Antibody

$262.68 - $460.68

Specifications

Antigen Foxp3
Clone PCH101
Host Species Rat
Gene Alias Forkhead Box P3, Scurfin, JM2, Treg
Applications Flow Cytometry (Intracellular Staining),Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011868 Life Technologies
LS13477680
25μg Each for $262.68
5011869 Life Technologies
LS13477682
100μg Each for $460.68
Specifications

Description

eBioscience offers a panel of monoclonal antibodies to different epitopes of human Foxp3, providing useful tools for investigating the complete expression pattern of Foxp3 at the protein level, and discerning the precise subsets of Foxp3+ cells.

The PCH101 antibody reacts with the amino terminus of human foxp3 protein also known as FORKHEAD BOX P3, SCURFIN, and JM2; cross reactivity of this antibody to other proteins has not been determined. Foxp3, a 49-55kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in ‘scurfy’ (sf) mice. Constitutive high expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg cells), and ectopic expression of foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

Intracellular staining of human peripheral blood mononuclear cells (PBMCs) with PCH101 antibody using the anti-human Foxp3 Staining Set and protocol reveals approximately 0.5-4% of lymphocytes staining, with the majority of staining occurring in the CD25bright population. This is subject to donor variability.

PCH101 crossreacts with rhesus, chimpanzee and cynomolgus. We recommend the use of CD4 (OKT4, cat. 11-0048, or RPA-T4, cat. 11-0049, depending on the species) and CD25 (BC96, cat. 17-0259).
Description & Specifications

Specifications

Foxp3
Rat
Flow Cytometry (Intracellular Staining),Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Western Blot
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
PCH101
Forkhead Box P3, Scurfin, JM2, Treg
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer