Please login to your online account to display your discounted pricing

Foxp3 Mouse anti-Human, eFluor 450, Clone: 236A/E7, eBioscience™

Mouse Monoclonal Antibody

$220.40 - $462.55

Specifications

Antigen Foxp3
Clone 236A/E7
Host Species Mouse
Gene Alias Forkhead Box P3, Scurfin, JM2
Species Reactivity Human, Non-human primate, Rhesus Ma
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016360

 
life technologies
LS48477741
25 tests Each for $220.40

501129020

 
life technologies
LS48477742
100 tests Each for $462.55
Description & Specifications

Specifications

Antigen Foxp3
Clone 236A/E7
Host Species Mouse
Gene Alias Forkhead Box P3, Scurfin, JM2
Species Reactivity Human, Non-human primate, Rhesus Ma
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The 236A/E7 antibody reacts with human foxp3 protein also known as FORKHEAD BOX P3, SCURFIN, and JM2; cross reactivity of this antibody to other proteins has not been determined. Foxp3, a 49-55kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in ‘scurfy’ (sf) mice. Constitutive high expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg cells), and ectopic expression of foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

Intracellular staining and flow cytometric analysis of freshly isolated human peripheral blood mononuclear cells (PBMCs) with the 236A/E7 antibody using the Foxp3 Staining Buffer Set (cat. 00-5523) and protocol reveals staining of the CD4+CD25bright population.

The epitope from 236A/E7 is different from that of PCH101 (cat. 72-5776).