missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
FOXP3, clone: 236A/E7, eBioscience™
Mouse Monoclonal Antibody
Manufacturer: Life Technologies LS14477782
Description
FOXP3 is a member of the forkhead/winged-helix family of transcriptional regulators and is highly conserved across mammals. FOXP3 is essential for normal immune homeostasis. FOXP3 is stably and constitutively expressed at a high level in CD25 + CD4 positive regulatory T cells, a low level in CD4-positive/CD25-negative cells, and is absent in CD4-negative/CD8-positive T cells. FOXP3 may be a master regulatory gene and a more specific marker of regulatory T cells than other T cells.
Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered. FOXP3 (Forkhead box protein 3) is a member of the forkhead/winged-helix family of transcriptional regulators, highly conserved across mammals, and essential for normal immune homeostasis. FOXP3 is 381 amino acids long, stably and constitutively expressed at a high level in CD25 + CD4 positive regulatory T cells, a low level in CD4-positive/CD25-negative cells, and is absent in CD4-negative/CD8-positive T cells. FOXP3 may be a master regulatory gene, and a more specific marker of regulatory T cells. Defects in the gene encoding FOXP3 protein cause the scurfy phenotype in mice. In humans FOXP3 defects play a role in IPEX syndrome (immune dysfunction, polyendocrinopathy, enteropathy, X-linked syndrome), also known as X-linked autoimmunity-allergic dysregulation (XLAAD) syndrome. Transcript variants of FOXP3 encoding different isoforms have been identified. In human breast and colon cancer cells, expression of FOXP3 is regulated by p53 in response to the DNA damage.Specifications
Foxp3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9BZS1 | |
AIID, DIETER, Forkhead box P3, foxp3, IPEX, JM2, PIDX, SCURFIN, XPID | |
Mouse | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human, Non-human Primate, Rhesus Monkey |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
236A/E7 | |
Unconjugated | |
FOXP3 | |
Liquid | |
FOXP3 | |
IgG1, kappa | |
100 μg | |
4° C | |
Primary | |
50943, 574303 |
For Research Use Only.