missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
FOXI1, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002299B02P
Description
Sequence: MSSFDLPAPSPPRCSPQFPSIGQEPPEMNLYYENFFHPQGVPSPQRPSFEGGGEYGATPNPYLWFNGPTMTPPPYLPGPNASPFLPQAYGVQRPLLPSVSGLGGSDLGWLPIPSQEELMKLVRPPYSYSALIAMAIHGAPDKRLTLSQIYQYVADNFPFYNKSKAGWQNSIRHNLSLNDCFKKVPRDEDDPAYVSGGSPTSHPLVTPGLSPEPSDKTGQNSLTFNSFSPLTNLSNHSGGGDWANPMPTNMLSYGGSVLSQFSPHFYNSVNTSGVLYPREGTEVSpecifications
FOXI1 | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
NM_144769.1 | |
FOXI1 | |
FOXI1 (NP_658982.1, 1 a.a. ∼ 283 a.a) full-length human protein. | |
RUO | |
2299 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Immunofluorescence,Western Blot | |
Unconjugated | |
forkhead box I1 | |
FKH10/FKHL10/FREAC6/HFH3/MGC34197 | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody |