Please login to your online account to display your discounted pricing

FoxA2 Mouse anti-Human, eFluor 660, Clone: 3C10, eBioscience™

Mouse Monoclonal Antibody

$160.95 - $397.30

Specifications

Antigen FoxA2
Clone 3C10
Host Species Mouse
Gene Alias Hepatocyte nuclear factor 3 beta
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124367

 
life technologies
LS50477880
25μg Each for $160.95

501124368

 
life technologies
LS50477882
100μg Each for $397.30
Description & Specifications

Specifications

Antigen FoxA2
Clone 3C10
Host Species Mouse
Gene Alias Hepatocyte nuclear factor 3 beta
Species Reactivity Human
Applications Immunohistochemistry (Formalin/Paraffin)
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This 3C10 monoclonal antibody reacts with human FOXA2, which is also known as hepatocyte nuclear factor 3 beta (HNF3B). This 48kDa member of the Forkhead family of transcription factors is expressed in the liver, pancreas (islets and some acinar cells), lung, and intestine. During development, FOXA2 is necessary for lung bud and pancreatic beta cell formation. Recently, directed expression of FOXA2 in embryonic stem cells has been found to regulate the generation and differentiation of midbrain dopaminergic neurons. It has also been shown to regulate genes involved hepatic lipid metabolism, including alpha fetoprotein, albumin, and transthyretin, as well as Pdx1 in the pancreas. FOXA2 expression is negatively regulated by insulin and reports demonstrate a role for it in sporadic cases of maturity-onset diabetes of the young and the epithelial-to-mesenchymal transition in cancer.