Please login to your online account to display your discounted pricing

Four-Stroke Gas and Diesel Engines

Air-cooled, side-valve four-stroke engines for gas or diesel.

$137.70 - $176.85

Specifications

For Use With (Application) Air cooled, side valve four stroke engines
Material Aluminum and Unbreakable Plastic
View More Specs
Products
Catalog Number Mfr. No. Weight (English) Weight (Metric) Product Type Price Quantity    

S96561

 
fisher science education
PH0488
5.29 lb. 2400g Petrol engine model Each for $137.70

S96560

 
fisher science education
PH0492
5.11 lb. 2320g Diesel engine model Each for $176.85
Description & Specifications

Specifications

For Use With (Application) Air cooled, side valve four stroke engines
Material Aluminum and Unbreakable Plastic

Gas Engine

  • Operation of valves clearly evident
  • Cams driven by a gear train from rear of main crank
  • Simulated spark plug uses a small lamp to indicate the firing point
  • Screw terminals provided for connection to 3V battery
  • Cross section of carburetor is shown

Diesel Engine
  • Chain-driven overhead camshaft
  • All functioning components (camshaft, rock-arms, tappets, etc.) demonstrated
  • Functioning fuel-injection system shown
  • Ignition represented by means of a miniature bulb

Mounted on base.