Please login to your online account to display your discounted pricing

American Educational Products Fossil Identification Lab


Uncover actual fossils more than 500 million years old.

Manufacturer: american educational products  3770

 View more versions of this product

Catalog No. S53500

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) 15 Students
For Use With (Application) Uncover actual fossils more than 500 million years old
Grade 5 to 12
Includes 15 fossils bearing rocks with 4 fossils each, 15 wash bottles, with tips and caps, 15 brushes, 15 Plastic magnifying Glasses, 15 Wooden digging tools, fossil card deck, 15 student guides, 15 geologic time mini-charts, 1 teacher guide
Type Fossil Identification Lab