missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Foss™ Tecator™ Kjeltec™ 8400 Analyzer Unit
The ultimate solution for automation in the routine Kjeldahl laboratory
Manufacturer: Foss North America 60063882
Includes: Tanks with level sensors for alkali/water/receiver solution/waste;, 100 and 250mL tubes; Combi-adapter for 100, 250, 400 and 750mL tubes.
Description
Automatic distillation and approved colorimetric titration for analysis of Kjeldahl nitrogen/protein, ammonium-nitrogen, TKN, Direct Distillation (DD), and volatile acids/bases.
Standard Features
- Automatic distillation procedure includes sample dilution, alkali and receiver addition, distillation, titration, calculation, reporting and tube draining
- Optional 20 or 60 place sample changer allows for fully unattended operation
- Complete PC control of all registration and reporting through optional data management software
- Alarms for reagent availability
- Accurate bellow pumps; high-precision refillable, interchangeable burette
- Intelligent cooling water control adjusts the flow according to temperature minimizing water consumption and costs
- Distillate temperature sensor automatically shuts down unit if risk exists of inaccurate results
- Built-in variable steam generator for deionized or tap water
- Alkali/water/receiver/waste tanks with level sensors
- Surfaces exposed to reagents are made of corrosion-free, chemical-resistant dyed plastic materials
- Self cleaning titration vessel
Safety Features
- Sensors for: 1) tube in place, 2) automatic safety door, 3) reagent level, 4) water level and 5) overpressure for steam generator
- SAfE™ (Steam Addition for Equilibration) technology improves safety by incorporating its own mixing procedure, reducing exothermic reaction between alkali and acid and making manual predilution of samples unnecessary
- A fast tube draining system removes the need for handling hot reagents after distillation
- Safety door and drip tray can both be easily removed for cleaning
Specifications
- Analysis time: 3.5 min. at 30mg nitrogen (6.5 min. at 200mg nitrogen)
- Distillation capacity: approximately 40mL/min
- Measuring range: 0.1—200mg nitrogen
- Reproducibility: ±1.0% relative (including the digestion step)
- Recovery is greater than 99.5% at nitrogen levels between 1 and 200mg
Order Info
Dimensions: 48L x 58W x 69cm H. Net Weight: 77 1/4 lb. (35kg).
Compliance
Officially approved colorimetric titration system (AOAC, EPA, DIN, ISO) simplifies validation. GLP Routines are supported by Recovery Test process and documentation of all events during analysis.