Please login to your online account to display your discounted pricing
Foss™ Kjeldahl Digestion Systems
Digestion units, with integrated programmable controllers, can be combined with a range of accessories to match individual needs
Manufacturer: foss north america  60062135
 View more versions of this product
Catalog No. TC36-102-0872
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Product Type Basic model digestion system
Voltage 230V
Electrical Requirements 230V 50/60Hz
For Use With (Equipment) 20 tubes
Quantity Each
Includes Digestion unit, tube rack with integrated heat shields, auto lift models include lift and integrated controller, auto rack models include rack and integrated controller
Hertz 50/60Hz