Please login to your online account to display your discounted pricing
Foss™ Kjeldahl Digestion Systems
Digestion units can be combined with a range of accessories to match individual needs. Auto models come include integrated programmable controllers.
Manufacturer: foss north america  60062134
 View more versions of this product
Catalog No. TC36-102-0870
  • / Each
Description & Specifications
Specifications
Product Type Basic model digestion system
Hertz 50/60Hz
Voltage 110V
Electrical Requirements 110V 50/60Hz
For Use With (Equipment) 8 tubes
Includes Digestion unit, tube rack with integrated heat shields, auto lift models include lift and integrated controller, auto rack models include rack and integrated controller
Quantity Each