Please login to your online account to display your discounted pricing
Foss™ Hydrocaps For Soxtec™ and Soxhlet Extraction Systems
For use in Hydrotec when final extraction is on Soxhlet, Soxtec™ 2050 or 2055 extraction systems.
$251.84 - $869.47
Specifications
For Use With (Application) For final extraction process
For Use With (Equipment) For Soxhlet, Soxtec™ 2050 or 2055 extraction system
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
TC60053554
foss™
60053554
240/Set Each for $869.47
TC60053553
foss north america
60053553
60/Set Each for $251.84
Description & Specifications
Specifications
For Use With (Application) For final extraction process
For Use With (Equipment) For Soxhlet, Soxtec™ 2050 or 2055 extraction system
For use in Hydrotec when final extraction is on Soxhlet, Soxtec 2050 or 2055 extraction systems.