Please login to your online account to display your discounted pricing

Foss™ Accessories for Soxtec™ Fat Extraction Systems: Butyl Seals


For use with Soxtec™ 2055 system and discontinued systems

Manufacturer: foss north america  10002517*

 View more versions of this product

Catalog No. TC1000-2517

  • / Pack of 6 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Soxtec™ unit HT 6, HT 12, 2043
Type Butyl Seal
Material Butyl Seal
Color Black
For Use With (Application) Fat extraction