missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
FOSB, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002354B01P
Description
Sequence: MPGSFVPTVTAITTSQDLQWLVQPTLISSMAQSQGQPLASQPPVVDPYDMPGTSYSTPGMSGYSSGGASGSGGPSTSGTTSGPGPARPARARPRRPREETLTPEEEEKRRVRRERNKLAAAKCRNRRRELTDRLQAETDQLEEEKAELESEIAELQKEKERLEFVLVAHKPGCKIPYEEGPGPGPLAEVRDLPGSAPAKEDGFSWLLPPPPPPPLPFQTSQDAPPNLTASLFTHSEVQVLGDPFPVVNPSYTSSFVLTCPEVSAFAGAQRTSGSDQPSDPLNSPSLLALSpecifications
FOSB | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
BC040197 | |
FOSB | |
FOSB (AAH40197.1, 1 a.a. ∼ 289 a.a) full-length human protein. | |
RUO | |
2354 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
FBJ murine osteosarcoma viral oncogene homolog B | |
AP-1/DKFZp686C0818/G0S3/GOS3/GOSB/MGC42291 | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody |