Please login to your online account to display your discounted pricing

Forskolin, Tocris


Adenylyl cyclase activator

Quantity 10mg

Manufacturer: r & d systems  RDS109910

 View more versions of this product

Catalog No. 109910

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 410.51
Quantity 10mg
CAS 66575-29-9
Chemical Name or Material Forskolin
Assay Percent Range >98%
Molecular Formula C22H34O7
Synonym [3R-(3α,4aβ,5β,6β,6aα,10α,10aβ,10bα)]-5-(Acetyloxy)-3-ethenyldodecahydro-6,10,10b-trihydroxy-3,4a,7,7,10a-penta
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material Forskolin
Assay Percent Range >98%
Molecular Formula C22H34O7
Formula Weight 410.51
Synonym [3R-(3α,4aβ,5β,6β,6aα,10α,10aβ,10bα)]-5-(Acetyloxy)-3-ethenyldodecahydro-6,10,10b-trihydroxy-3,4a,7,7,10a-penta
Accessories

Desiccate at -20°C