Please login to your online account to display your discounted pricing

Forskolin (From Coleus Forskohlii), Fisher BioReagents


Quantity 10mg
Packaging Amber Glass

Manufacturer: fisher scientific company  BP252010

 View more versions of this product

Catalog No. BP252010

  • / Each
  • / Case of 5 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 410.5
Melting Point 228°C
Quantity 10mg
Merck Index 15, 2460
Physical Form Solid
Molecular Formula C22 H34O7
Color Off-white
CAS 66575-29-9
Recommended Storage RT, desiccate
ChemAlert Storage Symbol Gray
Name Note From Coleus forskohlii
Packaging Amber Glass
Identification Pass Test
Chemical Name or Material Forskolin
Source Coleus Forskohlii
Synonym 7β-acetoxy-8,13-epoxy-1α,6β,9α-trihydroxy-labd-14-ene-11-one

Specifications

Chemical Name or Material Forskolin
Name Note From Coleus forskohlii
CAS 66575-29-9
Physical Form Solid
Molecular Formula C22 H34O7
Formula Weight 410.5
Synonym 7β-acetoxy-8,13-epoxy-1α,6β,9α-trihydroxy-labd-14-ene-11-one
Accessories
 

RT, desiccate

SDS & Safety Documents