missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Forskolin (From Coleus Forskohlii), Fisher BioReagents

Manufacturer:  Fisher BioReagents BP252025

 View more versions of this product

Catalog No. BP252025




Description

Description

Forskolin is a molecular biology reagent. Forskolin functions as an antihypertensive and vasodilator. Adenylcyclase activator.
Specifications

Specifications

Forskolin
66575-29-9
228°C
C22H34O7
15, 2460
25mg
CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O
410.507
CHEBI:42471
Solid
From Coleus forskohlii
White
Pass Test
Amber Glass
boforsin, coleonol, colforsin, colforsina, colforsina spanish, colforsine, colforsine french, colforsinum, colforsinum latin, forskolin
OHCQJHSOBUTRHG-KGGHGJDLSA-N
[(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate
47936
410.5

Chemical Identifiers

66575-29-9
410.507
boforsin, coleonol, colforsin, colforsina, colforsina spanish, colforsine, colforsine french, colforsinum, colforsinum latin, forskolin
CHEBI:42471
CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O
C22H34O7
OHCQJHSOBUTRHG-KGGHGJDLSA-N
47936
[(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate
Safety and Handling

Safety and Handling

Recommended Storage : RT, desiccate

SDS Documents
Product Certifications
Promotions

Promotions