missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Formylmethyl)triphenylphosphonium Chloride, TCI America™

Manufacturer:  TCI America F033125G

 View more versions of this product

Catalog No. F033125G



Specifications

Specifications

(Formylmethyl)triphenylphosphonium Chloride
25g
C20H18ClOP
formylmethyl triphenylphosphonium chloride, 2-oxoethyl triphenylphosphonium chloride, 2-triphenylphosphino ethanal, chloride, 2-oxoethyl triphenylphosphanium chloride, formylmethyl triphenylphosphonium chloride, acmc-1b4xm, 2-oxoethyltriphenylphosphonium chloride, formylmethyl triphenylphosphoniumchloride, 2-oxoethyl triphenyl phosphanium chloride, 2-oxoethyl triphenyl phosphonium chloride
C1=CC=C(C=C1)[P+](CC=O)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-]
340.787
340.79
≥98.0% (T)
Undesignated
62942-43-2
MFCD00012003
RVEJRPJGKXTQIF-UHFFFAOYSA-M
2-oxoethyl(triphenyl)phosphanium;chloride
2733863
Crystal-Powder at 20°C

Chemical Identifiers

62942-43-2
340.787
RVEJRPJGKXTQIF-UHFFFAOYSA-M
2733863
C1=CC=C(C=C1)[P+](CC=O)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-]
C20H18ClOP
MFCD00012003
formylmethyl triphenylphosphonium chloride, 2-oxoethyl triphenylphosphonium chloride, 2-triphenylphosphino ethanal, chloride, 2-oxoethyl triphenylphosphanium chloride, formylmethyl triphenylphosphonium chloride, acmc-1b4xm, 2-oxoethyltriphenylphosphonium chloride, formylmethyl triphenylphosphoniumchloride, 2-oxoethyl triphenyl phosphanium chloride, 2-oxoethyl triphenyl phosphonium chloride
2-oxoethyl(triphenyl)phosphanium;chloride
Safety and Handling

Safety and Handling

TSCA : No

Documents