Please login to your online account to display your discounted pricing

Alfa Aesar™ (Formylmethyl)triphenylphosphonium chloride, 98+%

$41.42 - $281.69

Chemical Identifiers

Chemical Name or Material (Formylmethyl)triphenylphosphonium chloride
CAS 62942-43-2
Molecular Formula OHCCH2P(C6H5)3Cl
Formula Weight 340.79
MDL Number MFCD00012003
Beilstein 4060813
Synonym 2-Oxoethyltriphenylphosphomium chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1573309

  SDS
alfa aesar™
A1573309
10g Each for $41.42

AAA1573318

  SDS
alfa aesar™
A1573318
50g Each for $155.47

AAA1573322

  SDS
alfa aesar™
A1573322
100g Each for $281.69
Description & Specifications

Specifications

Formula Weight 340.79
Chemical Name or Material (Formylmethyl)triphenylphosphonium chloride
Assay ≥98%
Molecular Formula OHCCH2P(C6H5)3Cl
MDL Number MFCD00012003
CAS 62942-43-2
Beilstein 4060813
Synonym 2-Oxoethyltriphenylphosphomium chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈210°C (dec.)
Quantity 10g

Applications
(Formylmethyl)triphenylphosphonium Chloride is a useful Wittig reagent. Used as a reactant for Hydroazidation reactions in preparation of a-azido alcohols, Preparation of prodrug (2-hydroxyamino-vinyl)-triphenyl-phosphonium (HVTP) inhibitor of peroxidase activity and apoptosis and Intramolecular [4 + 3] cycloaddition reactions.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool dry place in tightly closed container. Store away from oxidizing agent and water/moisture.

Safety and Handling

Danger

GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.