missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (Formylmethyl)triphenylphosphonium chloride, 98+%

Manufacturer:  Alfa Aesar™ A1573309

 View more versions of this product

Catalog No. AAA1573309



Description

Description

Applications
(Formylmethyl)triphenylphosphonium Chloride is a useful Wittig reagent. Used as a reactant for Hydroazidation reactions in preparation of a-azido alcohols, Preparation of prodrug (2-hydroxyamino-vinyl)-triphenyl-phosphonium (HVTP) inhibitor of peroxidase activity and apoptosis and Intramolecular [4 + 3] cycloaddition reactions.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool dry place in tightly closed container. Store away from oxidizing agent and water/moisture.
Specifications

Specifications

(Formylmethyl)triphenylphosphonium chloride
10g
C20H18ClOP
4060813
RVEJRPJGKXTQIF-UHFFFAOYSA-M
2-oxoethyl(triphenyl)phosphanium;chloride
2733863
≥98%
~210°C (decomposition)
62942-43-2
MFCD00012003
formylmethyl triphenylphosphonium chloride, 2-oxoethyl triphenylphosphonium chloride, 2-triphenylphosphino ethanal, chloride, 2-oxoethyl triphenylphosphanium chloride, formylmethyl triphenylphosphonium chloride, acmc-1b4xm, 2-oxoethyltriphenylphosphonium chloride, formylmethyl triphenylphosphoniumchloride, 2-oxoethyl triphenyl phosphanium chloride, 2-oxoethyl triphenyl phosphonium chloride
C1=CC=C(C=C1)[P+](CC=O)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-]
340.787
340.79

Chemical Identifiers

62942-43-2
340.787
RVEJRPJGKXTQIF-UHFFFAOYSA-M
2733863
C1=CC=C(C=C1)[P+](CC=O)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-]
C20H18ClOP
MFCD00012003
formylmethyl triphenylphosphonium chloride, 2-oxoethyl triphenylphosphonium chloride, 2-triphenylphosphino ethanal, chloride, 2-oxoethyl triphenylphosphanium chloride, formylmethyl triphenylphosphonium chloride, acmc-1b4xm, 2-oxoethyltriphenylphosphonium chloride, formylmethyl triphenylphosphoniumchloride, 2-oxoethyl triphenyl phosphanium chloride, 2-oxoethyl triphenyl phosphonium chloride
2-oxoethyl(triphenyl)phosphanium;chloride
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents