Please login to your online account to display your discounted pricing

Formoterol hemifumarate, Tocris

Potent and selective β2 agonist

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material Formoterol hemifumarate
Assay Percent Range >99%
Molecular Formula C19H24N2O4·1/2C4H4O4
Formula Weight 402.45
Synonym (±)-(R, R)-N-[2-Hydroxy-5-[1-hydroxy-2-[[2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl] formamide hemifumarate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

144810

 
r & d systems
RDS144810
10mg Each for $181.19

144850

 
r & d systems
RDS144850
50mg Each for $762.58
Description & Specifications

Specifications

Formula Weight 402.45
Quantity 10mg
Chemical Name or Material Formoterol hemifumarate
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C19H24N2O4·1/2C4H4O4
CAS 43229-80-7
Synonym (±)-(R, R)-N-[2-Hydroxy-5-[1-hydroxy-2-[[2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl] formamide hemifumarate
Safety and Handling

Store at Room Temperature