Please login to your online account to display your discounted pricing

Formic Acid, Reagent ACS 88+%, ACROS Organics™


Quantity 10L
Packaging Plastic Jerrycan

Manufacturer: thermo fisher scientific  AC410770100

 View more versions of this product

Catalog No. AC410770100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Assay ≥88.0%
Physical Form Liquid
Formula Weight 46.02
Quantity 10L
Color Clear
Packaging Plastic Jerrycan
Density 1.2200g/mL
Fieser 01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177
Residue after Evaporation 0.002% max.
Iron (Fe) 5ppm max.
Heavy Metals (as Pb) 5ppm max.
Sulfite Passes test
Linear Formula HCO2H
Assay Percent Range 88%
Dilution Test Passes test
Health Hazard 1 Danger
Free Acid 0.4% max.
Infrared Spectrum Authentic
Chlorine (Cl) 10ppm max.
CAS 64-18-6
Ammonium 50ppm max.
Sulfate 20ppm max.
Refractive Index 1.3694 to 1.3714 (20°C, 589nm)
Chemical Name or Material Formic acid
Molecular Formula CH2O2

Specifications

Chemical Name or Material Formic acid
CAS 64-18-6
Purity Grade ACS Reagent Grade
Assay ≥88.0%
Assay Percent Range 88%
Molecular Formula CH2O2
Linear Formula HCO2H
Formula Weight 46.02
Accessories

Danger

SDS & Safety Documents