missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formic Acid, LC-MS Ultra, Honeywell™ Fluka™

$402.06 - $402.06

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044843 View Documents Honeywell Chemicals
142652ML
2mL Ampoule Each for $402.06
 
Specifications

Chemical Identifiers

64-18-6
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
284
formic acid
CH2O2
MFCD00003297
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
CHEBI:30751
C(=O)O

Specifications

64-18-6
CH2O2
MFCD00003297
1209246
1.22g/mL at 25°C
C(=O)O
46.025
CHEBI:30751
0.57
1004°F
approx.98% (T)
44.8mmHg (20°C)
Liquid
Colorless
2mL
Formic Acid
Eluent additive for UHPLC-MS
HCOOH
UN1779
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
BDAGIHXWWSANSR-UHFFFAOYSA-N
formic acid
284
46.03g/mol
LC-MS Ultra
Ampoule
n20/D 1.370(lit.)
1.6 (vs air)
4°C
100.4°C
49°C (120.2°F)
Safety and Handling

Safety and Handling

P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P233

H226-H302-H314-H331

DOTInformation : 3(8)/PGII

EINECSNumber : 200-579-1

RTECSNumber : LQ4900000