Learn More
Formic acid hydrazide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1490122
Description
Formic acid hydrazide is used as a precursor in the preparation of 1,2,4-triazole derivatives. It is also used in the synthesis of 6-N-formylamino-12,13-dihydro-1,11-dihydroxy-13-(beta-D-glucopyranosyl)-5H-indolo[2,3-a]-pyrrolo[3,4-c]carbazole-5,7(6H)-dione, which is an anticancer agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFormic acid hydrazide is used as a precursor in the preparation of 1,2,4-triazole derivatives. It is also used in the synthesis of 6-N-formylamino-12,13-dihydro-1,11-dihydroxy-13-(beta-D-glucopyranosyl)-5H-indolo[2,3-a]-pyrrolo[3,4-c]carbazole-5,7(6H)-dione, which is an anticancer agent.
Solubility
Soluble in water, ethanol and acetone.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
Specifications
Formic acid hydrazide | |
55°C to 59°C | |
CH4N2O | |
100 g | |
Hygroscopic | |
Soluble in water,ethanol and acetone. | |
C(=O)NN | |
60.056 | |
60.06 |
624-84-0 | |
>110°C (230°F) | |
MFCD00007589 | |
635759 | |
formylhydrazine, formic acid hydrazide, formic hydrazide, hydrazinecarboxaldehyde, formhydrazide, n-formylhydrazine, carbazaldehyde, formylhydrazide, hydrazine, formyl, formal hydrazine | |
XZBIXDPGRMLSTC-UHFFFAOYSA-N | |
formohydrazide | |
12229 | |
98% |
Chemical Identifiers
624-84-0 | |
60.056 | |
XZBIXDPGRMLSTC-UHFFFAOYSA-N | |
12229 | |
C(=O)NN |
CH4N2O | |
MFCD00007589 | |
formylhydrazine, formic acid hydrazide, formic hydrazide, hydrazinecarboxaldehyde, formhydrazide, n-formylhydrazine, carbazaldehyde, formylhydrazide, hydrazine, formyl, formal hydrazine | |
formohydrazide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 210-867-9
RTECSNumber : LQ8615000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only