missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formic acid-d2, for NMR, +99% atom D, 95% solution in D2O, ACROS Organics™

$645.68 - $645.68

Chemical Identifiers

CAS 920-42-3
Molecular Formula CH2O2
Molecular Weight (g/mol) 48.037
MDL Number MFCD00037363
InChI Key BDAGIHXWWSANSR-PFUFQJKNSA-N
Synonym 2h formic 2 acid, 2h2 formic acid, dcood, formic acid-d2, formic acid-d2, 95 wt. % in d2o, 98 atom % d, formic-d acid-d 6ci,7ci,8ci,9ci, formic-d acid-d, 95 wt% in deuterium oxide
PubChem CID 123092
IUPAC Name deuterio deuterioformate
SMILES C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC233400100
SDS View Documents
Thermo Fisher Scientific
233400100
10g Glass bottle Each for $645.68
 
Description

Description

For NMR
Specifications

Chemical Identifiers

920-42-3
48.037
BDAGIHXWWSANSR-PFUFQJKNSA-N
123092
C(=O)O
CH2O2
MFCD00037363
2h formic 2 acid, 2h2 formic acid, dcood, formic acid-d2, formic acid-d2, 95 wt. % in d2o, 98 atom % d, formic-d acid-d 6ci,7ci,8ci,9ci, formic-d acid-d, 95 wt% in deuterium oxide
deuterio deuterioformate

Specifications

1.2700g/mL
50°C
1.27
Colorless
10g
CH2O2
MFCD00037363
Solubility in water: miscible.
C(=O)O
48.037
48.04
≥94.0%
Formicacid-d2, 99+ atom % D (95% in D2O)
99.0% min.
Glass bottle
101.0°C
5.9°C
920-42-3
DCOOD
2h formic 2 acid, 2h2 formic acid, dcood, formic acid-d2, formic acid-d2, 95 wt. % in d2o, 98 atom % d, formic-d acid-d 6ci,7ci,8ci,9ci, formic-d acid-d, 95 wt% in deuterium oxide
BDAGIHXWWSANSR-PFUFQJKNSA-N
deuterio deuterioformate
123092
Liquid
NMR
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-057-3