Learn More
Formic acid, 99%, for analysis, ACROS Organics™
Formic acid, 99%, CH2O2, CAS Number-64-18-6, acide formique, aminic acid, bilorin, collo-bueglatt, formira, formisoton, formylic acid, hydrogen carboxylic acid, methanoic acid, myrmicyl, 2.5L, Vapor Pressure: 44mbar at 20 deg.C, 02, 8, 101.0 deg.C, 0.1ppm max., CHEBI:30751, 5ppm max., Undesignated
Manufacturer: Thermo Fisher Scientific 270480250
Description
Formic acid, 99%, CH2O2, CAS Number-64-18-6, acide formique, aminic acid, bilorin, collo-bueglatt, formira, formisoton, formylic acid, hydrogen carboxylic acid, methanoic acid, myrmicyl, 2.5L, Vapor Pressure: 44mbar at 20 deg.C, 02, 8, 101.0 deg.C, 0.1ppm max., CHEBI:30751, 5ppm max., Undesignated
For analysisSpecifications
Formic acid | |
Vapor Pressure: 44mbar at 20°C | |
0.1ppm max. | |
HCO2H | |
5ppm max. | |
01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177 | |
acide formique, aminic acid, bilorin, collo-bueglatt, formira, formisoton, formylic acid, hydrogen carboxylic acid, methanoic acid, myrmicyl | |
1.2200g/mL | |
0.1ppm max. | |
formic acid | |
284 | |
46.02 | |
99% | |
Authentic | |
0.1ppm max. | |
0.005% max. | |
1.3700 to 1.3720 | |
50ppm max. | |
8.0°C | |
101.0°C | |
50°C |
64-18-6 | |
99% | |
CH2O2 | |
MFCD00003297 | |
02, 8 | |
15, 4269 | |
Solubility in water: miscible. Other solubilities: soluble in alcohol, ether and glycerine | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
C(=O)O | |
46.025 | |
CHEBI:30751 | |
Liquid | |
Analysis | |
2ppm max. | |
Passes Test | |
Plastic Bottle | |
1.22 | |
20ppm max. | |
Undesignated | |
25mL |
Chemical Identifiers
64-18-6 | |
46.025 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
284 | |
formic acid |
CH2O2 | |
MFCD00003297 | |
acide formique, aminic acid, bilorin, collo-bueglatt, formira, formisoton, formylic acid, hydrogen carboxylic acid, methanoic acid, myrmicyl | |
CHEBI:30751 | |
C(=O)O |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger