Learn More
Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 423755000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Formic acid | |
8.0°C | |
101.0°C | |
CH2O2 | |
MFCD00003297 | |
01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
formic acid | |
284 | |
46.02 | |
ACS Reagent | |
500 g | |
Authentic | |
Liquid | |
0.003% max. | |
44mbar at 20°C |
96+% | |
64-18-6 | |
97% | |
HCO2H | |
02, 8 | |
15, 4269 | |
Solubility in water: miscible. Other solubilities: soluble in alcohol, ether and glycerine | |
C(=O)O | |
46.025 | |
CHEBI:30751 | |
97% | |
1.2200g/mL | |
50°C | |
Plastic bottle | |
1.3700 to 1.3720 | |
1.22 |
Chemical Identifiers
64-18-6 | |
46.025 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
284 | |
formic acid |
CH2O2 | |
MFCD00003297 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
CHEBI:30751 | |
C(=O)O |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 200-579-1
RTECSNumber : LQ4900000
TSCA : TSCA
RUO – Research Use Only