Please login to your online account to display your discounted pricing

Formic Acid, 90%, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$336.82 - $1,347.03

Chemical Identifiers

Chemical Name or Material Formic Acid
CAS 64-18-6
Name Note Polymer Characterization Solvent
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula HCOOH
Formula Weight 46.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002909

 
avantor performance
012905
Glass Bottle 2.5L Each for $1,347.03

02002910

 
avantor performance
012901
Glass Bottle 500mL Each for $336.82
Description & Specifications

Specifications

Formula Weight 46.03
Chemical Name or Material Formic Acid
Assay 90%
CAS 64-18-6
Specific Gravity 1L = 1.20kg
Quantity 2.5L
Flash Point 50.00°C
Name Note Polymer Characterization Solvent
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula HCOOH
Packaging Glass Bottle