Please login to your online account to display your discounted pricing

Formic Acid, 88%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$239.74 - $573.88

Chemical Identifiers

Chemical Name or Material Formic Acid
CAS 64-18-6
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula HCOOH
Formula Weight 46.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002907

 
avantor performance
012805
Glass Bottle 2.5L Each for $573.88

02002908

 
avantor performance
012801
Glass Bottle 500mL Each for $239.74
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 46.03
Chemical Name or Material Formic Acid
Molecular Formula HCOOH
Specific Gravity 1L = 1.20kg
Quantity 2.5L
Flash Point 50.00°C
Assay 88%
CAS 64-18-6
Packaging Glass Bottle