missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formanilide, TCI America™

Manufacturer:  TCI America F0047500G

 View more versions of this product

Catalog No. F0047500G



Specifications

Specifications

Formanilide
Undesignated
500g
C7H7NO
formanilide, formamide, n-phenyl, n-formylaniline, phenyl formamide, formamidobenzene, formylaniline, carbanilaldehyde, aniline, n-formyl, phenylformamide, n-phenyl-formamide
C1=CC=C(C=C1)NC=O
121.139
CHEBI:42416
Crystal-Powder at 20°C
271°C
48°C
103-70-8
MFCD00003276
DYDNPESBYVVLBO-UHFFFAOYSA-N
N-phenylformamide
7671
121.14
≥99.0% (GC)

Chemical Identifiers

103-70-8
121.139
DYDNPESBYVVLBO-UHFFFAOYSA-N
7671
N-phenylformamide
C7H7NO
MFCD00003276
formanilide, formamide, n-phenyl, n-formylaniline, phenyl formamide, formamidobenzene, formylaniline, carbanilaldehyde, aniline, n-formyl, phenylformamide, n-phenyl-formamide
CHEBI:42416
C1=CC=C(C=C1)NC=O
Safety and Handling

Safety and Handling

RTECSNumber : BY1780000

TSCA : Yes

Recommended Storage : Refrigerator

Documents