missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Formanilide, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 158421000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Formanilide | |
103-70-8 | |
Beige-Gray to White | |
271°C | |
Authentic | |
Plastic bottle | |
HCONHC6H5 | |
100 g | |
1.14 | |
formanilide, formamide, n-phenyl, n-formylaniline, phenyl formamide, formamidobenzene, formylaniline, carbanilaldehyde, aniline, n-formyl, phenylformamide, n-phenyl-formamide | |
DYDNPESBYVVLBO-UHFFFAOYSA-N | |
N-phenylformamide | |
7671 | |
121.14 | |
Crystals |
99+% | |
47°C to 49°C | |
1.1400g/mL | |
>112°C | |
99% min. (GC) | |
C7H7NO | |
MFCD00003276 | |
12, 230 | |
15, 4266 | |
Solubility in water: 25.4g/L (20 c) | |
C1=CC=C(C=C1)NC=O | |
121.14 | |
CHEBI:42416 | |
99+% |
Chemical Identifiers
103-70-8 | |
DYDNPESBYVVLBO-UHFFFAOYSA-N | |
7671 | |
N-phenylformamide |
MFCD00003276 | |
formanilide, formamide, n-phenyl, n-formylaniline, phenyl formamide, formamidobenzene, formylaniline, carbanilaldehyde, aniline, n-formyl, phenylformamide, n-phenyl-formamide | |
CHEBI:42416 | |
C1=CC=C(C=C1)NC=O |
Safety and Handling
EINECSNumber : 203-136-0
RTECSNumber : BY1780000
TSCA : TSCA
RUO – Research Use Only