missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formanilide, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 158420050

 View more versions of this product

Catalog No. AC158420050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Formanilide
103-70-8
Beige-Gray to White
271°C
Authentic
Glass bottle
HCONHC6H5
5 g
1.14
formanilide, formamide, n-phenyl, n-formylaniline, phenyl formamide, formamidobenzene, formylaniline, carbanilaldehyde, aniline, n-formyl, phenylformamide, n-phenyl-formamide
DYDNPESBYVVLBO-UHFFFAOYSA-N
N-phenylformamide
7671
121.14
Crystals
99+%
47°C to 49°C
1.1400g/mL
>112°C
99% min. (GC)
C7H7NO
MFCD00003276
12, 230
15, 4266
Solubility in water: 25.4g/L (20 c)
C1=CC=C(C=C1)NC=O
121.14
CHEBI:42416
99+%

Chemical Identifiers

103-70-8
DYDNPESBYVVLBO-UHFFFAOYSA-N
7671
N-phenylformamide
MFCD00003276
formanilide, formamide, n-phenyl, n-formylaniline, phenyl formamide, formamidobenzene, formylaniline, carbanilaldehyde, aniline, n-formyl, phenylformamide, n-phenyl-formamide
CHEBI:42416
C1=CC=C(C=C1)NC=O
Safety and Handling

Safety and Handling

EINECSNumber : 203-136-0

RTECSNumber : BY1780000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only