missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formamidinesulfinic Acid, TCI America™

Manufacturer:  TCI America F0115500G

 View more versions of this product

Catalog No. F0115500G



Specifications

Specifications

Formamidinesulfinic Acid
126°C
1758-73-2
MFCD00002397
thiourea dioxide, formamidinesulfinic acid, amino imino methanesulfinic acid, formamidine sulfinic acid, manofast, thiourea s,s-dioxide, aimsa, methanesulfinic acid, aminoimino, carbamimidoylsulfinic acid, methenesulfinic acid, aminoimino
C(=N)(N)S(=O)O
108.115
108.12
≥93.0% (T)
White
500g
CH4N2O2S
3341
FYOWZTWVYZOZSI-UHFFFAOYSA-N
amino(imino)methanesulfinic acid
61274
Crystal-Powder at 20°C

Chemical Identifiers

1758-73-2
108.115
FYOWZTWVYZOZSI-UHFFFAOYSA-N
61274
C(=N)(N)S(=O)O
CH4N2O2S
MFCD00002397
thiourea dioxide, formamidinesulfinic acid, amino imino methanesulfinic acid, formamidine sulfinic acid, manofast, thiourea s,s-dioxide, aimsa, methanesulfinic acid, aminoimino, carbamimidoylsulfinic acid, methenesulfinic acid, aminoimino
amino(imino)methanesulfinic acid
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2873

RTECSNumber : PB0372500

TSCA : Yes

Documents