Learn More
Formamidinesulfinic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 119721000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Formamidinesulfinic acid | |
1758-73-2 | |
White | |
99% | |
CH4N2O2S | |
MFCD00002397 | |
03, I, 36 | |
thiourea dioxide, formamidinesulfinic acid, amino imino methanesulfinic acid, formamidine sulfinic acid, manofast, thiourea s,s-dioxide, aimsa, methanesulfinic acid, aminoimino, carbamimidoylsulfinic acid, methenesulfinic acid, aminoimino | |
FYOWZTWVYZOZSI-UHFFFAOYSA-N | |
amino(imino)methanesulfinic acid | |
61274 | |
99% |
99% | |
126.0°C | |
Authentic | |
Plastic bottle | |
H2NC(=NH)SO2H | |
100 g | |
04,506; 05,668; 06,586 | |
Solubility in water: 30g/L (20°C). Other solubilities: insoluble in organic solvents | |
C(=N)(N)S(=O)O | |
108.11 | |
108.11 | |
Crystalline Powder |
Chemical Identifiers
1758-73-2 | |
108.11 | |
FYOWZTWVYZOZSI-UHFFFAOYSA-N | |
61274 | |
C(=N)(N)S(=O)O |
CH4N2O2S | |
MFCD00002397 | |
thiourea dioxide, formamidinesulfinic acid, amino imino methanesulfinic acid, formamidine sulfinic acid, manofast, thiourea s,s-dioxide, aimsa, methanesulfinic acid, aminoimino, carbamimidoylsulfinic acid, methenesulfinic acid, aminoimino | |
amino(imino)methanesulfinic acid |
Safety and Handling
GHS H Statement
Self-heating in large quantities; may catch fire.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
May cause damage t
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep cool.
Protect from sunlight.
Avoid breathing dust/fume/ga
GHS Signal Word: Danger
EINECSNumber : 217-157-8
TSCA : TSCA
RUO – Research Use Only