missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Formamidinesulfinic acid, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 119725000
Specifications
Formamidinesulfinic acid | |
Plastic bottle | |
White | |
500g | |
1758-73-2 | |
CH4N2O2S | |
MFCD00002397 | |
04,506; 05,668; 06,586 | |
Solubility in water: 30g/L (20°C). Other solubilities: insoluble in organic solvents | |
C(=N)(N)S(=O)O | |
108.11 | |
108.11 | |
99% |
Authentic | |
0.5% max. | |
126.0°C | |
99% | |
99% | |
H2NC(=NH)SO2H | |
03, I, 36 | |
aimsa, amino imino methanesulfinic acid, carbamimidoylsulfinic acid, formamidine sulfinic acid, formamidinesulfinic acid, manofast, methanesulfinic acid, aminoimino, methenesulfinic acid, aminoimino, thiourea dioxide, thiourea s,s-dioxide | |
FYOWZTWVYZOZSI-UHFFFAOYSA-N | |
amino(imino)methanesulfinic acid | |
61274 | |
Crystalline Powder |
Chemical Identifiers
1758-73-2 | |
108.11 | |
FYOWZTWVYZOZSI-UHFFFAOYSA-N | |
61274 | |
C(=N)(N)S(=O)O |
CH4N2O2S | |
MFCD00002397 | |
aimsa, amino imino methanesulfinic acid, carbamimidoylsulfinic acid, formamidine sulfinic acid, formamidinesulfinic acid, manofast, methanesulfinic acid, aminoimino, methenesulfinic acid, aminoimino, thiourea dioxide, thiourea s,s-dioxide | |
amino(imino)methanesulfinic acid |
Safety and Handling
GHS H Statement
Self-heating in large quantities; may catch fire.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
May cause damage t
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep cool.
Protect from sunlight.
Avoid breathing dust/fume/ga
GHS Signal Word: Danger
EINECSNumber : 217-157-8
TSCA : TSCA