missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Formamidine Hydrochloride 97.0+%, TCI America™
Supplier: TCI America F01035G
Specifications
Formamidine Hydrochloride | |
85°C | |
CH4ClN2 | |
5 g | |
OEJWQRAWUVWBIE-UHFFFAOYSA-N | |
methanimidamide hydrochloridyl | |
10313058 | |
≥97.0% (T) |
6313-33-3 | |
White | |
MFCD00012865 | |
formamidine hydrochloride, formimidamide hydrochloride, formamidinium chloride, methanimidamide, monohydrochloride, formamidine hcl, formimidamide hcl, formamidine hcl salt, imidoformamide hydrochloride, methanimidamide hydrochloride | |
[Cl].NC=N | |
79.51 | |
80.52 | |
Crystalline Powder |
Chemical Identifiers
6313-33-3 | |
79.51 | |
OEJWQRAWUVWBIE-UHFFFAOYSA-N | |
10313058 | |
[Cl].NC=N |
CH4ClN2 | |
MFCD00012865 | |
formamidine hydrochloride, formimidamide hydrochloride, formamidinium chloride, methanimidamide, monohydrochloride, formamidine hcl, formimidamide hcl, formamidine hcl salt, imidoformamide hydrochloride, methanimidamide hydrochloride | |
methanimidamide hydrochloridyl |
Safety and Handling
TSCA : No