missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formamidine Disulfide Dihydrochloride, TCI America™

Manufacturer:  TCI America F004625G

 View more versions of this product

Catalog No. F004625G



Specifications

Specifications

Formamidine Disulfide Dihydrochloride
25g
C2H8Cl2N4S2
formamidine disulfide dihydrochloride, formamdine disulfide dihydrochloride, dithiourea dihydrochloride, c2h6n4s2.2hcl, carbamimidoyldisulfanylmethanimidamide dihydrochloride, formamidine disulfide dihydrochloride disulfide dihydrochloride
C(=N)(N)SSC(=N)N.Cl.Cl
223.134
223.13
≥98.0% (N,T)
White-Yellow
14807-75-1
MFCD00012586
BFJQSCVWXZOXGK-UHFFFAOYSA-N
carbamimidoylsulfanyl carbamimidothioate;dihydrochloride
24802135
Crystal-Powder at 20°C

Chemical Identifiers

14807-75-1
223.134
BFJQSCVWXZOXGK-UHFFFAOYSA-N
24802135
C(=N)(N)SSC(=N)N.Cl.Cl
C2H8Cl2N4S2
MFCD00012586
formamidine disulfide dihydrochloride, formamdine disulfide dihydrochloride, dithiourea dihydrochloride, c2h6n4s2.2hcl, carbamimidoyldisulfanylmethanimidamide dihydrochloride, formamidine disulfide dihydrochloride disulfide dihydrochloride
carbamimidoylsulfanyl carbamimidothioate;dihydrochloride
Safety and Handling

Safety and Handling

RTECSNumber : LQ4570000

TSCA : Yes

Documents