Learn More
Formamidine acetate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1115836
Description
Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFormamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.
Solubility
Soluble in water and hot alcohol.
Notes
Hygroscopic. Moisture sensitive. Incompatible with oxidizing agents.
Specifications
Formamidine acetate | |
∼158°C (decomposition) | |
8 | |
MFCD00012866 | |
3563488 | |
formamidine acetate, methanimidamide, monoacetate, formamidinium acetate, acetic acid; methanimidamide, formamidine acetate salt, formamidine, monoacetate, formimidamide acetate, imidoformamide acetate, acetic acid; formamidine, methanimidamide monoacetate | |
XPOLVIIHTDKJRY-UHFFFAOYSA-N | |
acetic acid;methanimidamide | |
160693 | |
99% |
3473-63-0 | |
1.124 | |
C3H8N2O2 | |
500 g | |
Hygroscopic | |
Soluble in water and hot alcohol. | |
CC(=O)O.C(=N)N | |
104.109 | |
104.11 |
Chemical Identifiers
3473-63-0 | |
104.109 | |
XPOLVIIHTDKJRY-UHFFFAOYSA-N | |
160693 | |
CC(=O)O.C(=N)N |
C3H8N2O2 | |
MFCD00012866 | |
formamidine acetate, methanimidamide, monoacetate, formamidinium acetate, acetic acid; methanimidamide, formamidine acetate salt, formamidine, monoacetate, formimidamide acetate, imidoformamide acetate, acetic acid; formamidine, methanimidamide monoacetate | |
acetic acid;methanimidamide |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
H315-H317-H319-H335
EINECSNumber : 222-442-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only