Please login to your online account to display your discounted pricing

Formamide, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

$105.05 - $218.38

Chemical Identifiers

Chemical Name or Material Formamide
CAS 75-12-7
Purity Grade Ultrapure Bioreagent
Molecular Formula HCONH2
Formula Weight 45.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002903

 
avantor performance
402800
Narrow Mouth Amber Glass Bottle 100mL Each for $105.05

02002905

 
avantor performance
402801
Narrow Mouth Amber Glass Bottle 500mL Each for $218.38
Description & Specifications

Specifications

Formula Weight 45.04
Specific Gravity 1L = 1.13kg
Purity Grade Ultrapure Bioreagent
Quantity 100mL
Molecular Formula HCONH2
CAS 75-12-7
Chemical Name or Material Formamide
Packaging Narrow Mouth Amber Glass Bottle
Flash Point 154.00°C

    For denaturing nucleic acids prior to analysis; For Laboratory,Research,or Manufacturing Use