Please login to your online account to display your discounted pricing

Alfa Aesar™ Formamide, Ultra Pure, 99.5% min


Quantity 250mL

Manufacturer: alfa aesar™  J67206AK

 View more versions of this product

Catalog No. AAJ67206AK

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 45.04
Physical Form Liquid
Chemical Name or Material Formamide
CAS 75-12-7
MDL Number MFCD00007941
Health Hazard 2 GHS H Statement
H360
May damage fertility or the unborn child.
Odor Ammonia-like
Refractive Index 1.4475
Assay ≥99.5%
Health Hazard 1 Danger
Name Note Ultra Pure
Density 1.134g/mL
Quantity 250mL
RTECS Number LQ0525000
Beilstein 505995
TSCA TSCA
Sensitivity Hygroscopic
Color Clear, colorless
Boiling Point 210°C
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 2°C

Specifications

Chemical Name or Material Formamide
CAS 75-12-7
Name Note Ultra Pure
Formula Weight 45.04
MDL Number MFCD00007941
Beilstein 505995

Applications
Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.

Solubility
Soluble in water (20 mg/ml ), methanol, ethanol and acetone. Slightly soluble in ether.

Notes
Hygroscopic. Store at room temperature. Store away from water or moisture.

Accessories

Danger

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360
May damage fertility or the unborn child.

SDS & Safety Documents