Learn More
Formamide, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 044066AY
Description
Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Formamide | |
75-12-7 | |
1.134 g/mL | |
4 x 500 mL | |
Ammonia-like | |
1.4475 | |
HCONH2 | |
505995 | |
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
formamide | |
713 | |
45.04 | |
Spectrophotometric |
2°C | |
Colorless | |
210°C | |
154°C (310°F) | |
Liquid | |
CH3NO | |
MFCD00007941 | |
14,4237 | |
Soluble in water (20mg/ml ),methanol,ethanol and acetone. Slightly soluble in ether. | |
NC=O | |
45.04 | |
CHEBI:48431 | |
≥99% |
Chemical Identifiers
75-12-7 | |
45.04 | |
ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
713 | |
formamide |
CH3NO | |
MFCD00007941 | |
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
CHEBI:48431 | |
NC=O |
Safety and Handling
GHS H Statement
H360
May damage fertility or the unborn child.
P201-P202-P260-P281-P308+P313-P501c
H351-H360FD-H373
EINECSNumber : 200-842-0
RTECSNumber : LQ0525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only